ENA447957471 Enamine

N-(5-chloro-2-methoxyphenyl)-4-methylpiperidine-1-carboxamide

  • MDL number MFCD04294054
  • Cas Number : 60465-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207127
Availability details for
Quantity
  • Availability
  • Add to Cart