ENA447957472 Enamine

2-ethyl-1-[(3-methylphenyl)methyl]-1H-1,3-benzodiazole

  • MDL number MFCD03714169
  • Cas Number : 537018-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207128
Availability details for
Quantity
  • Availability
  • Add to Cart