ENA447957473 Enamine

4-phenoxy-1H-indole

  • MDL number MFCD11869062
  • Cas Number : 930790-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207138
Availability details for
Quantity
  • Availability
  • Add to Cart