ENA447957475 Enamine

3-{[4-(2-ethoxy-2-oxoethyl)-1,3-thiazol-2-yl]carbamoyl}prop-2-enoic acid

  • Cas Number : 327105-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205216
Availability details for
Quantity
  • Availability
  • Add to Cart