ENA447957485 Enamine

tert-butyl 3-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

  • MDL number MFCD18383202
  • Cas Number : 869852-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207251
Availability details for
Quantity
  • Availability
  • Add to Cart