ENA447957486 Enamine

2-[5-(trifluoromethyl)pyridin-2-yl]propanedinitrile

  • MDL number MFCD02713592
  • Cas Number : 377760-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207254
Availability details for
Quantity
  • Availability
  • Add to Cart