ENA447957492 Enamine

[2-amino-1-(furan-2-yl)ethyl](butan-2-yl)amine

  • MDL number MFCD21196468
  • Cas Number : 1498050-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207265
Availability details for
Quantity
  • Availability
  • Add to Cart