ENA447957493 Enamine

[2-amino-1-(furan-2-yl)ethyl](benzyl)amine

  • MDL number MFCD21226869
  • Cas Number : 128349-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207266
Availability details for
Quantity
  • Availability
  • Add to Cart