ENA447957501 Enamine

8-phenyl-1-oxa-3-azaspiro[4.5]dec-2-en-2-amine

  • MDL number MFCD21032397
  • Cas Number : 1483028-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205113
Availability details for
Quantity
  • Availability
  • Add to Cart