ENA447957503 Enamine

2-amino-5,5-dimethyl-4,5-dihydro-1,3-thiazol-4-one

  • MDL number MFCD19204133
  • Cas Number : 4695-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207280
Availability details for
Quantity
  • Availability
  • Add to Cart