ENA447957506 Enamine

6-amino-2,3-dihydro-1H-indene-5-sulfonic acid

  • MDL number MFCD24623875
  • Cas Number : 36125-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207290
Availability details for
Quantity
  • Availability
  • Add to Cart