ENA447957512 Enamine

N,N,4-trimethyl-1,3-thiazol-2-amine

  • MDL number MFCD13179624
  • Cas Number : 6142-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207298
Availability details for
Quantity
  • Availability
  • Add to Cart