ENA447957514 Enamine

1-(2-fluoropyridin-4-yl)ethan-1-one

  • MDL number MFCD09263924
  • Cas Number : 111887-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37-39
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207301
Availability details for
Quantity
  • Availability
  • Add to Cart