ENA447957515 Enamine

methyl 2-(3,4-dichlorophenyl)-3-hydroxypropanoate

  • MDL number MFCD23134077
  • Cas Number : 1254365-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207302
Availability details for
Quantity
  • Availability
  • Add to Cart