ENA447957517 Enamine

2,6-dichloro-3-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD11109540
  • Cas Number : 276702-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207307
Availability details for
Quantity
  • Availability
  • Add to Cart