ENA447957518 Enamine

methyl 3,5-dichloro-2-(chlorosulfonyl)benzoate

  • MDL number MFCD11650112
  • Cas Number : 1098387-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207308
Availability details for
Quantity
  • Availability
  • Add to Cart