ENA447957519 Enamine

methyl 3-[3-(3-methoxy-3-oxopropyl)phenyl]propanoate

  • MDL number MFCD01109873
  • Cas Number : 6221-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207310
Availability details for
Quantity
  • Availability
  • Add to Cart