ENA447957533 Enamine

2-(4-chlorophenyl)-5-methoxypentan-1-amine

  • MDL number MFCD21925655
  • Cas Number : 1542705-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207341
Availability details for
Quantity
  • Availability
  • Add to Cart