ENA447957535 Enamine

2-{[4-(trifluoromethyl)phenyl]methyl}butanoic acid

  • MDL number MFCD16503009
  • Cas Number : 1269529-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205619
Availability details for
Quantity
  • Availability
  • Add to Cart