ENA447957545 Enamine

4-phenyl-N-(piperidin-3-yl)butanamide hydrochloride

  • MDL number MFCD28384030
  • Cas Number : 1803604-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207370
Availability details for
Quantity
  • Availability
  • Add to Cart