ENA447957549 Enamine

1-(5-chloro-2,4-dimethoxyphenyl)-3,3-dimethylurea

  • MDL number MFCD26969321
  • Cas Number : 1708195-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207374
Availability details for
Quantity
  • Availability
  • Add to Cart