ENA447957553 Enamine

1-cyclopropyl-2-(2-methylphenyl)ethan-1-one

  • MDL number MFCD11934673
  • Cas Number : 56594-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205639
Availability details for
Quantity
  • Availability
  • Add to Cart