ENA447957555 Enamine

2-(6-sulfanylhexyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD22905249
  • Cas Number : 85847-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207409
Availability details for
Quantity
  • Availability
  • Add to Cart