ENA447957560 Enamine

(3-cyano-4-hydroxyphenyl)boronic acid

  • MDL number MFCD18394168
  • Cas Number : 1637208-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207449
Availability details for
Quantity
  • Availability
  • Add to Cart