ENA447957562 Enamine

[2-amino-1-(4-chlorophenyl)ethyl](methyl)(3,3,3-trifluoropropyl)amine

  • MDL number MFCD21023737
  • Cas Number : 1466767-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205645
Availability details for
Quantity
  • Availability
  • Add to Cart