ENA447957563 Enamine

2-(1-aminocyclopentyl)ethan-1-ol

  • MDL number MFCD19207613
  • Cas Number : 896734-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207452
Availability details for
Quantity
  • Availability
  • Add to Cart