ENA447957565 Enamine

methyl 2-{2-[(tert-butoxy)methyl]phenyl}acetate

  • MDL number MFCD23659590
  • Cas Number : 1552448-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207455
Availability details for
Quantity
  • Availability
  • Add to Cart