ENA447957568 Enamine

1-(4-ethylphenyl)-3-methylcyclohexan-1-ol

  • MDL number MFCD16793768
  • Cas Number : 1248098-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207459
Availability details for
Quantity
  • Availability
  • Add to Cart