ENA447957570 Enamine

N-benzyl-N-methylpropanamide

  • MDL number MFCD05660035
  • Cas Number : 34317-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205648
Availability details for
Quantity
  • Availability
  • Add to Cart