ENA447957578 Enamine

2-(4-chlorophenyl)-3-hydroxypropanoic acid

  • MDL number MFCD24479915
  • Cas Number : 16864-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207475
Availability details for
Quantity
  • Availability
  • Add to Cart