ENA447957583 Enamine

ethyl 2-{1,1,3-trioxo-2H,3H-1lambda6-[1,2]thiazolo[5,4-b]pyridin-2-yl}acetate

  • MDL number MFCD26419046
  • Cas Number : 174468-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207488
Availability details for
Quantity
  • Availability
  • Add to Cart