ENA447957584 Enamine

N-(1,1-dioxo-1lambda6-thiolan-3-yl)-N-methylcarbamoyl chloride

  • MDL number MFCD23790162
  • Cas Number : 1536007-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205664
Availability details for
Quantity
  • Availability
  • Add to Cart