ENA447957587 Enamine

[1-(cyanomethyl)cyclopropyl]methanesulfonyl chloride

  • MDL number MFCD21244836
  • Cas Number : 1466482-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207495
Availability details for
Quantity
  • Availability
  • Add to Cart