ENA447957595 Enamine

5-(chlorosulfonyl)-2-methylbenzoyl chloride

  • MDL number MFCD28397465
  • Cas Number : 1426437-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207533
Availability details for
Quantity
  • Availability
  • Add to Cart