ENA447957609 Enamine

propyl[2-(pyrrolidin-1-yl)ethyl]amine

  • MDL number MFCD11159861
  • Cas Number : 138356-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207564
Availability details for
Quantity
  • Availability
  • Add to Cart