ENA447957611 Enamine

4-(ethoxycarbonyl)oxane-4-carboxylic acid

  • MDL number MFCD24485511
  • Cas Number : 108630-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207566
Availability details for
Quantity
  • Availability
  • Add to Cart