ENA447957613 Enamine

7-chloro-4-methyl-1,3-benzothiazol-2-amine

  • MDL number MFCD04971843
  • Cas Number : 78584-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-207
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207568
Availability details for
Quantity
  • Availability
  • Add to Cart