ENA447957614 Enamine

azetidine-1-sulfonyl chloride

  • MDL number MFCD19200214
  • Cas Number : 639519-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207569
Availability details for
Quantity
  • Availability
  • Add to Cart