ENA447957617 Enamine

3-methoxy-4-(3-methoxypropoxy)aniline

  • MDL number MFCD16684117
  • Cas Number : 1249721-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207572
Availability details for
Quantity
  • Availability
  • Add to Cart