ENA447957620 Enamine

2-bromo-5-phenyl-1,3-oxazole

  • MDL number MFCD12828094
  • Cas Number : 129053-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207575
Availability details for
Quantity
  • Availability
  • Add to Cart