ENA447957621 Enamine

3-(2-ethylphenoxy)propanoic acid

  • MDL number MFCD09704596
  • Cas Number : 938125-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207578
Availability details for
Quantity
  • Availability
  • Add to Cart