ENA447957626 Enamine

1-[4-(3-methoxypropoxy)phenyl]methanamine

  • MDL number MFCD11620912
  • Cas Number : 1094783-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207585
Availability details for
Quantity
  • Availability
  • Add to Cart