ENA447957631 Enamine

2-(pyrrolidin-1-yl)-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD25576481
  • Cas Number : 865470-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207593
Availability details for
Quantity
  • Availability
  • Add to Cart