ENA447957636 Enamine

3-(pyridin-2-yl)-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD19685618
  • Cas Number : 56041-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 285-289
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207599
Availability details for
Quantity
  • Availability
  • Add to Cart