ENA447957637 Enamine

N-[(2-chlorophenyl)methyl]-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD06192753
  • Cas Number : 120161-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-184
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205731
Availability details for
Quantity
  • Availability
  • Add to Cart