ENA447957645 Enamine

4-(hexa-2,4-dienamido)benzoic acid

  • Cas Number : 945155-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205736
Availability details for
Quantity
  • Availability
  • Add to Cart