ENA447957647 Enamine

(2R)-2-[(1-cyanocyclopentyl)formamido]propanoic acid

  • MDL number MFCD28383932
  • Cas Number : 1604394-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205737
Availability details for
Quantity
  • Availability
  • Add to Cart