ENA447957653 Enamine

methyl 2-methoxy-2-phenylacetate

  • MDL number MFCD00040999
  • Cas Number : 3558-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205740
Availability details for
Quantity
  • Availability
  • Add to Cart