ENA447957655 Enamine

7-methoxy-1,2,3,4-tetrahydroquinolin-4-one

  • MDL number MFCD00796999
  • Cas Number : 879-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207674
Availability details for
Quantity
  • Availability
  • Add to Cart