ENA447957656 Enamine

2,7-dichloroimidazo[1,2-a]pyridine-3-carbaldehyde

  • MDL number MFCD22076956
  • Cas Number : 1536845-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207678
Availability details for
Quantity
  • Availability
  • Add to Cart